Skip to Content
MilliporeSigma

03485

N-Acetyl-3-O-β-D-galactopyranosyl-β-D-galactosamine methyl glycoside

≥98.0% (TLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H27NO11
CAS Number:
Molecular Weight:
397.37
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5485654
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98.0% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

storage temp.

2-8°C

SMILES string

CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@H]1NC(C)=O

InChI

1S/C15H27NO11/c1-5(19)16-8-13(10(21)7(4-18)25-14(8)24-2)27-15-12(23)11(22)9(20)6(3-17)26-15/h6-15,17-18,20-23H,3-4H2,1-2H3,(H,16,19)/t6-,7-,8-,9+,10+,11+,12-,13-,14-,15+/m1/s1

InChI key

JPIHIQIYWZWIPU-NNPCJPQPSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library