Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D8184

Millipore

Diagnostic sensitivity test agar

BioChemika, suitable for microbiology

Synonym(s):

D.S.T. Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

pH

7.4±0.2 (25 °C)

application(s)

microbiology

Application

Used as an antibiotic sensitivity testing medium.

Components

Componentsg/L
Proteose peptone 10.00
Veal infusion solids 10.00
Dextrose2.00
Sodium chloride3.00
Disodium phosphate2.00
Sodium acetate1.00
Adenine sulphate0.01
Guanine hydrochloride0.01
Uracil0.01
Xanthine0.01
Aneurine0.00002
Agar15.00

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bechtle, R.M., et al.
Antibiotics and Chemotherapy, 8(12), 599-599 (1958)
A standard culture medium for general bacteriology.
J H MARSHALL et al.
The Journal of hygiene, 58, 367-372 (1960-12-01)
WHO
Organic Process Research & Development, 210 (1961)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service