Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIEH04265CBA

Rieke Metals, Inc.

4-(3-CHLORO-4-FLUOROPHENYL)THIOPHENOL

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
238.71
MDL number:

Iupac Name

3'-chloro-4'-fluoro[1,1'-biphenyl]-4-thiol

Inchi Code

1S/C12H8ClFS/c13-11-7-9(3-6-12(11)14)8-1-4-10(15)5-2-8/h1-7,15H

InChI key

OKEUEXJWKBPJAB-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Product Link

https://riekemetals.com/products-services/catnum/NV05745

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service