Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIE515935261

Rieke Metals, Inc.

4-Bromo-3-methoxyphenylzinc iodide 0.5 M in Tetrahydrofuran

0.5 M in Tetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
378.32

Iupac Name

(4-bromo-3-methoxyphenyl)zinc(II) iodide

Inchi Code

1S/C7H6BrO.HI.Zn/c1-9-7-5-3-2-4-6(7)8;;/h2,4-5H,1H3;1H;/q;;+1/p-1

InChI key

YUENAYHAFKOXRW-UHFFFAOYSA-M

Country of Origin

US

Physical Form

Liquid

Product Link

https://riekemetals.com/products-services/catnum/2358

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service