Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIE240869916

Rieke Metals, Inc.

5-Chloro-2-methoxyphenylzinc bromide 0.5 M in Tetrahydrofuran

0.5 M in Tetrahydrofuran

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$438.92
100 ML
$715.13

Vendor SKU: 2296-50ML

$438.92


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 ML
$438.92
100 ML
$715.13

About This Item

Molecular Weight:
286.87
MDL number:

Vendor SKU: 2296-50ML

$438.92


Ships in 14 days - from Aldrich Partner.

Iupac Name

bromo(5-chloro-2-methoxyphenyl)zinc

Inchi Code

1S/C7H6ClO.BrH.Zn/c1-9-7-4-2-6(8)3-5-7;;/h2-4H,1H3;1H;/q;;+1/p-1

InChI key

CEIPZJDHUITUFD-UHFFFAOYSA-M

Country of Origin

US

Physical Form

Liquid

Product Link

https://riekemetals.com/products-services/catnum/2296

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service