RIE156320882
Rieke Metals, Inc.
3,3',4-Trifluorobenzophenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(3,4-difluorophenyl)(3-fluorophenyl)methanone
Inchi Code
1S/C13H7F3O/c14-10-3-1-2-8(6-10)13(17)9-4-5-11(15)12(16)7-9/h1-7H
InChI key
KNZKTGFPHVAIRB-UHFFFAOYSA-N
Purity
97%
Country of Origin
US
Product Link
https://riekemetals.com/products-services/catnum/5204-41
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service