Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIE156320590

Rieke Metals, Inc.

3-Acetoxy-4'-cyanobenzophenone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
265.27
MDL number:

Iupac Name

3-(4-cyanobenzoyl)phenyl acetate

Inchi Code

1S/C16H11NO3/c1-11(18)20-15-4-2-3-14(9-15)16(19)13-7-5-12(10-17)6-8-13/h2-9H,1H3

InChI key

RQHBBYSMZAPZJF-UHFFFAOYSA-N

Purity

90%

Country of Origin

US

Product Link

https://riekemetals.com/products-services/catnum/5176-25

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service