Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIE156319488

Rieke Metals, Inc.

cis-2-(3-fluorobenzoyl)cyclohexane-1-carboxylic acid

All chiral compounds contain a mixture of enantiomers i.e. not chirally pure.

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$828.03
5 G
$2,030.67
10 G
$3,551.25

Vendor SKU: 18509-22-1G

$828.03


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
$828.03
5 G
$2,030.67
10 G
$3,551.25

About This Item

CAS Number:
Molecular Weight:
250.27
MDL number:

Vendor SKU: 18509-22-1G

$828.03


Ships in 14 days - from Aldrich Partner.

Iupac Name

(1R,2S)-2-(3-fluorobenzoyl)cyclohexanecarboxylic acid

Inchi Code

1S/C14H15FO3/c15-10-5-3-4-9(8-10)13(16)11-6-1-2-7-12(11)14(17)18/h3-5,8,11-12H,1-2,6-7H2,(H,17,18)/t11-,12+/m0/s1

InChI key

YJIFMHRCDUOGST-NWDGAFQWSA-N

Purity

95%

Country of Origin

US

Product Link

https://riekemetals.com/products-services/catnum/18509-22

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service