Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIE156319154

Rieke Metals, Inc.

2-bromo-4'carboethoxybenzophenone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$375.99
2 G
$689.79
5 G
$1,238.58

Vendor SKU: 5011-1G

$375.99


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 G
$375.99
2 G
$689.79
5 G
$1,238.58

About This Item

CAS Number:
Molecular Weight:
333.18
MDL number:

Vendor SKU: 5011-1G

$375.99


Ships in 14 days - from Aldrich Partner.

Iupac Name

ethyl 4-(2-bromobenzoyl)benzoate

Inchi Code

1S/C16H13BrO3/c1-2-20-16(19)12-9-7-11(8-10-12)15(18)13-5-3-4-6-14(13)17/h3-10H,2H2,1H3

InChI key

KJEVRARDFQLTST-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

White solid

Product Link

https://riekemetals.com/products-services/catnum/5011

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service