Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RIE156319024

Rieke Metals, Inc.

2,5-Dimethoxyphenylzinc iodide 0.5 M in Tetrahydrofuran

0.5 M in Tetrahydrofuran

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$570.09
100 ML
$891.71

Vendor SKU: 2205-50ML

$570.09


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 ML
$570.09
100 ML
$891.71

About This Item

CAS Number:
Molecular Weight:
329.45
MDL number:

Vendor SKU: 2205-50ML

$570.09


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

(2,5-dimethoxyphenyl)(iodo)zinc

Inchi Code

1S/C8H9O2.HI.Zn/c1-9-7-3-5-8(10-2)6-4-7;;/h3-5H,1-2H3;1H;/q;;+1/p-1

Inchi Key

JJTGBAXIFVYPNT-UHFFFAOYSA-M

Country of Origin

US

Physical Form

Liquid

Product Link

https://riekemetals.com/products-services/catnum/2205

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service