PRIH93ED8261
Princeton BioMolecular Research, Inc.
2-isopropyl-4-(4-nitrophenyl)-1,3-thiazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
248.31
MDL number:
Iupac Name
2-isopropyl-4-(4-nitrophenyl)-1,3-thiazole
Inchi Code
1S/C12H12N2O2S/c1-8(2)12-13-11(7-17-12)9-3-5-10(6-4-9)14(15)16/h3-8H,1-2H3
InChI key
OTBJIEUPODKWMZ-UHFFFAOYSA-N
Purity
95%
Country of Origin
UA
Product Link
http://store.princetonbio.com/?product_id=PBMR362934
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service