Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PRIH93ED821C

Princeton BioMolecular Research, Inc.

methyl 3-aminothieno[2,3-b]quinoline-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
258.3
MDL number:

Iupac Name

methyl 3-aminothieno[2,3-b]quinoline-2-carboxylate

Inchi Code

1S/C13H10N2O2S/c1-17-13(16)11-10(14)8-6-7-4-2-3-5-9(7)15-12(8)18-11/h2-6H,14H2,1H3

InChI key

VPTFPLOAHPWQAK-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

Solid

Product Link

http://store.princetonbio.com/?product_id=PBMR362865

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service