Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PRIH93ED811F

Princeton BioMolecular Research, Inc.

5H-chromeno[2,3-c]isoquinolin-5-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$439.67
250 MG
$571.59
500 MG
$743.07
1 G
$966.00

Vendor SKU: PBMR362612-100MG

$439.67


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$439.67
250 MG
$571.59
500 MG
$743.07
1 G
$966.00

About This Item

Molecular Weight:
247.25
MDL number:

Vendor SKU: PBMR362612-100MG

$439.67


Ships in 21 days - from Aldrich Partner.

Iupac Name

5H-chromeno[2,3-c]isoquinolin-5-one

Inchi Code

1S/C16H9NO2/c18-15-12-7-3-2-6-11(12)13-9-10-5-1-4-8-14(10)19-16(13)17-15/h1-9H

InChI key

JLKGOYDXVVKMFS-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

Solid

Product Link

http://store.princetonbio.com/?product_id=PBMR362612

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service