Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PRIH049B2FA2

Princeton BioMolecular Research, Inc.

sodium (2,5-dimethylphenyl)methanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Molecular Weight:
222.24
MDL number:

Iupac Name

sodium (2,5-dimethylphenyl)methanesulfonate

Inchi Code

1S/C9H12O3S.Na/c1-7-3-4-8(2)9(5-7)6-13(10,11)12;/h3-5H,6H2,1-2H3,(H,10,11,12);/q;+1/p-1

InChI key

YOXMKKPNRUCWDT-UHFFFAOYSA-M

Purity

95%

Country of Origin

UA

Physical Form

N/A

Product Link

http://store.princetonbio.com/?product_id=PBMR219028

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service