Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PRIH039FD6FF

Princeton BioMolecular Research, Inc.

3-Benzo[1,3]dioxol-5-yl-3-oxo-propionic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
222.2
MDL number:

Iupac Name

methyl 3-(1,3-benzodioxol-5-yl)-3-oxopropanoate

Inchi Code

1S/C11H10O5/c1-14-11(13)5-8(12)7-2-3-9-10(4-7)16-6-15-9/h2-4H,5-6H2,1H3

InChI key

QYNFBQXZTZKAEB-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Product Link

http://store.princetonbio.com/?product_id=PBMR269360

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service