Skip to Content
MilliporeSigma

ABC472

Anti-PRKCI Antibody/Protein kinase C iota

from rabbit, purified by affinity chromatography

Synonym(s):

Protein kinase C iota type, Atypical protein kinase C-lambda/iota, PRKC-lambda/iota, aPKC-lambda/iota, nPKC-iota

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human
Application:
ICC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRKCI(5584)

General description

PRKCI, or Protein kinase C iota, is an important calcium- and diacylglycerol-independent serine/ threonine-protein kinase that plays a general protective role against apoptotic stimuli, regulates NF-kappa-B activation, cell survival, cell differentiation and polarity, and contributes to the regulation of microtubule dynamics during development. PRKCI also plays roles in many other systems including neuronal survival and it helps organize the apical growth and polarization of epithelial cellular extensions. PRKCI is localized in cytoplasm, membranes, endosomes, and nucleus. PRKCI is predominantly expressed in lung and brain but is also expressed at lower levels in many tissues including is pancreatic islets. PRKCI is also over expressed in many cancers including small cell lung cancers.
~66 kDa observed

Immunogen

Epitope: Protein Kinase Domain
KLH-conjugated linear peptide corresponding to the Protein Kinase Domain of human PRKCI/Protein kinase C iota.

Application

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected PRKCI/Protein kinase C iota in HeLa, A431, and NIH/3T3 cells.
Alexa Fluor is a registered trademark of Life Technologies.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-PRKCI Antibody/Protein kinase C iota is validated for use in Western Blotting and Immunocytochemistry for the detection of PRKCI/Protein kinase C iota.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected PRKCI/Protein kinase C iota in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library