Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

742

Sigma-Aldrich

Wright-Giemsa Stain

HARLECO® Polychromic stain for peripheral blood and bone marrow smears. Specifically formulated for staining malarial parasites.

Synonym(s):

Hematology Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

Quality Level

form

liquid

mp

19.85 °C

transition temp

flash point 12.2 °C

density

0.82 g/cm3

λmax

520-530 nm (Eosin)
645-655 nm (Azure)

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

Application

HARLECO® Polychromic stain for peripheral blood and bone marrow smears. Specifically formulated for staining malarial parasites.
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Polychromic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation. Specifically formulated for staining malarial parasites.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

54.0 °F

flash_point_c

12.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service