Skip to Content
MilliporeSigma

566331

SIRT2 Inhibitor II, AK-1

The SIRT2 Inhibitor II, AK-1, also referenced under CAS 330461-64-8, controls the biological activity of SIRT2. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

SIRT2 Inhibitor II, AK-1, 3-(azepan-1-ylsulfonyl)-N-(3-nitrophenyl)benzamide, SIRT2 Inhibitor II, AK-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H21N3O5S
CAS Number:
Molecular Weight:
403.45
UNSPSC Code:
12352200
NACRES:
NA.28
MDL number:
Assay:
>95% (HPLC)
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

>95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N3CCCCCC3)c1cc(ccc1)C(=O)Nc2cc(ccc2)[N+](=O)[O-]

InChI

1S/C19H21N3O5S/c23-19(20-16-8-6-9-17(14-16)22(24)25)15-7-5-10-18(13-15)28(26,27)21-11-3-1-2-4-12-21/h5-10,13-14H,1-4,11-12H2,(H,20,23)

InChI key

HAYBKCHPEBZNGW-UHFFFAOYSA-N

General description

A cell permeable, benzylsulfonamide that inhibits SIRT2 activity (IC50= 12.5 µM through enzymatic assay) by targeting the SIRT2 nicotinamide binding site, but may also target SIRT1 and SIRT3 (IC50 >40 µM) with much less potency. Western analysis illustrates that treatment with AK-1 (>25 µM) leads to an increase in acetylated tubulin. AK-1 (1 to 10 µM) is shown to rescue neuronal dysfunction in two in vivo models of Huntington′s Disease (HD) in Dropshila and Caenorhabditis elegans. It also demonstrates recovery of neuronal toxicity in a primary model of HD from 1µM to 4 µM similar to that of genetic SIRT2 inhibition, and also correlates with the negative regulation of sterol biosynthesis.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Luthi-Carter, R., et al. 2010. Proc. Natl. Acad. Sci.107, 7927.
Outeiro, TF., et al. 2007. Science317, 516.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library