162
Brilliant Green (C.I. 42040)
HARLECO® Bacterial stain
Synonym(s):
Brilliant Green, Malachite Green G, C. I. 42040
About This Item
Recommended Products
Quality Level
form
solid
pH
2.5 (25 °C, 10 g/L in H2O)
mp
210 °C (decomposition)
solubility
100 g/L
λmax
628-632 nm
application(s)
diagnostic assay manufacturing
hematology
histology
shipped in
ambient
storage temp.
15-25°C
SMILES string
[S](=O)(=O)([O-])[O-].[N+](=C1C=CC(=C(c3ccc(cc3)N(CC)CC)c2ccccc2)C=C1)(CC)CC.[H+]
InChI
1S/C27H33N2.H2O4S/c1-5-28(6-2)25-18-14-23(15-19-25)27(22-12-10-9-11-13-22)24-16-20-26(21-17-24)29(7-3)8-4;1-5(2,3)4/h9-21H,5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1
InChI key
NNBFNNNWANBMTI-UHFFFAOYSA-M
General description
162-12 25g Glass Bottle
Application
For In-Vitro Diagnostic Use.
For Professional Use.
Also used as a component of brilliant green bile media for the selective enrichment and enumeration of E. coli and other fecal coliform organisms in water, milk and other foodstuffs.
Analysis Note
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service