This buffer gives us a beautiful coloration and differenciation with the Wright-Giemsa Fucillo stain.
1217
Buffer, Phosphate pH 6.4
Harleco®, suitable for hematology
About This Item
Recommended Products
form
liquid
Quality Level
manufacturer/tradename
Harleco®
color
colorless
pH
6.4
mp
-0.1 °C
application(s)
diagnostic assay manufacturing
hematology
shipped in
ambient
storage temp.
10-30°C
General description
1217-78 1L Poly Pottle
Application
Legal Information
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service