MilliporeSigma
9502S-PP-CLEAR
MicroSolv
MicroSolv Polypropylene Vials
2 ml Vials, screw top, plastic. Translucent, 9-425 mm threads, 12x32 mm outer dimensions. For use as an autosampler vial. MicroSolv Brand. 100/PK.
A Trusted Marketplace Seller
Synonym(s):
Non-ionic
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
O.D.
12x32 mm
volume
2 mL, 2000 μL
General description
Our most popular plastic autosampler vial can used instead of glass vials for HPLC. These high volume (2ml) blow-molded, plastic vials are made with virgin polypropylene, have fill lines with graduations and a slightly concave bottom that is helpful in reducing residual volume.
Non-ionic, these PP vials can reduce or eliminate adsorption of many compounds including proteins, peptides and bases where adsorption is common with borosilicate glass.
Excellent choice for Ion Chromatography, CE, Bio-analysis, analysis of PFAS or simply replacing basic HPLC autosampler vials. Compatible with most modern autosamplers that accept 12x32mm vials.
Still not finding the right product?
Explore all of our products under MicroSolv Polypropylene Vials
Choose from one of the most recent versions:
Certificates of Analysis (COA)