Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAT390503266

Matrix Scientific

5-Amino-2-methylphenol sulfate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
221.23
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

sulfuric acid compound with 5-amino-2-methylphenol (1:1)

Inchi Code

1S/C7H9NO.H2O4S/c1-5-2-3-6(8)4-7(5)9;1-5(2,3)4/h2-4,9H,8H2,1H3;(H2,1,2,3,4)

InChI key

OOUDTDWQXPSATC-UHFFFAOYSA-N

storage temp.

Ambient

Purity

95%

Country of Origin

US

Product Link

https://www.matrixscientific.com/091933.html

pictograms

MSDS

https://www.matrixscientific.com/091933.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service