MAT321761414
Matrix Scientific
2,2-Dichloro-1-(4-chlorophenyl)ethanone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
128/10 Torr
mp
61-62
Iupac Name
2,2-dichloro-1-(4-chlorophenyl)ethanone
Inchi Code
1S/C8H5Cl3O/c9-6-3-1-5(2-4-6)7(12)8(10)11/h1-4,8H
InChI key
BZOFRNRRMMEHOB-UHFFFAOYSA-N
Country of Origin
US
Product Link
https://www.matrixscientific.com/059776.html
Boiling point
128/10 Torr
Melting Point
61-62
MSDS
https://www.matrixscientific.com/059776.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service