Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAT048697480

Matrix Scientific

4-(Perfluorocyclohexyl)butan-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$99.60

Vendor SKU: 012870-1G

$99.60


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
$99.60

About This Item

CAS Number:
Molecular Weight:
354.16
MDL number:

Vendor SKU: 012870-1G

$99.60


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

4-(1,2,2,3,3,4,4,5,5,6,6-undecafluorocyclohexyl)-1-butanol

Inchi Code

1S/C10H9F11O/c11-5(3-1-2-4-22)6(12,13)8(16,17)10(20,21)9(18,19)7(5,14)15/h22H,1-4H2

InChI key

BEMDKZLEDMTSTG-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Product Link

https://www.matrixscientific.com/012870.html

pictograms

MSDS

https://www.matrixscientific.com/012870.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service