MAT047026588
Matrix Scientific
5-Amino-1-isopropyl-1H-pyrazol-3-ol
Select a Size
Vendor SKU: 008282-500MG
$201.60
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
Vendor SKU: 008282-500MG
$201.60
Notify Me
Get notified when this item is ready to ship via email.
mp
>300
Iupac Name
5-amino-1-isopropyl-1H-pyrazol-3-ol
Inchi Code
1S/C6H11N3O/c1-4(2)9-5(7)3-6(10)8-9/h3-4H,7H2,1-2H3,(H,8,10)
InChI key
FHPXLYGNSXYUPE-UHFFFAOYSA-N
Country of Origin
Product Link
Melting Point
MSDS
https://www.matrixscientific.com/008282.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service