MAT047025560
Matrix Scientific
4-[(5-Bromopyrimidin-2-yl)thio]phenylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
107-109
Iupac Name
4-[(5-bromo-2-pyrimidinyl)sulfanyl]aniline
Inchi Code
1S/C10H8BrN3S/c11-7-5-13-10(14-6-7)15-9-3-1-8(12)2-4-9/h1-6H,12H2
InChI key
RXDXNLRTAYPJFH-UHFFFAOYSA-N
Country of Origin
US
Product Link
https://www.matrixscientific.com/007335.html
Melting Point
107-109
MSDS
https://www.matrixscientific.com/007335.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service