Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAT047021464

Matrix Scientific

3-Ethoxy-4-methoxyphenylacetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$60.00

Vendor SKU: 002561-1G

$60.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$60.00

About This Item

CAS Number:
Molecular Weight:
191.23
MDL number:

Vendor SKU: 002561-1G

$60.00


Ships in 7 days - from Aldrich Partner.

bp

151/2.5 Torr

mp

61

Iupac Name

(3-ethoxy-4-methoxyphenyl)acetonitrile

Inchi Code

1S/C11H13NO2/c1-3-14-11-8-9(6-7-12)4-5-10(11)13-2/h4-5,8H,3,6H2,1-2H3

InChI key

DULUKHADKMYILX-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Product Link

https://www.matrixscientific.com/002561.html

Boiling point

151/2.5 Torr

Melting Point

61

MSDS

https://www.matrixscientific.com/002561.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service