MAN458951301
Manchester Organics
2-Chloropyrimidine-5-carbonyl chloride
About This Item
Iupac Name
2-chloro-5-pyrimidinecarbonyl chloride
Inchi Code
1S/C5H2Cl2N2O/c6-4(10)3-1-8-5(7)9-2-3/h1-2H
InChI key
JUYQWJULYXCBAV-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service