Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LIFH942E9E78

Life Chemicals Inc.

2-methyl-3-[(piperidin-3-yl)methoxy]pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
207.28
MDL number:

Iupac Name

2-methyl-3-(piperidin-3-ylmethoxy)pyrazine

Inchi Code

1S/C11H17N3O/c1-9-11(14-6-5-13-9)15-8-10-3-2-4-12-7-10/h5-6,10,12H,2-4,7-8H2,1H3

InChI key

XWVSPZZXTHKUMS-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

oil

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service