LIF009175994
Life Chemicals Inc.
1-(5-Nitro-1H-benzimidazol-2-yl)ethanol
Select a Size
Vendor SKU: F0910-7378-250MG
$198.90
Availability
Select a Size
About This Item
Vendor SKU: F0910-7378-250MG
$198.90
Availability
Iupac Name
1-(5-nitro-1H-benzimidazol-2-yl)ethanol
Inchi Code
1S/C9H9N3O3/c1-5(13)9-10-7-3-2-6(12(14)15)4-8(7)11-9/h2-5,13H,1H3,(H,10,11)
InChI key
IVFGODDNHJTTBF-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service