Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY947335473

Key Organics / BIONET

12-oxapentacyclo[11.8.0.0²,¹¹.0³,?.0¹?,²¹]henicosa-1(13),2(11),3(8),4,6,9,14,16(21),17,19-decaene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
268.31
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

155 - 156

Iupac Name

dinaphtho[2,1-b:1',2'-d]furan

Inchi Code

1S/C20H12O/c1-3-7-15-13(5-1)9-11-17-19(15)20-16-8-4-2-6-14(16)10-12-18(20)21-17/h1-12H

InChI key

QIMOZEUDRUTIBS-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/NS-00075

Melting Point

155 - 156

pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary statements

MSDS

https://www.keyorganics.net/sds/NS-00075


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service