Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY465203281

Key Organics / BIONET

tricyclo[5.2.1.0²,?]dec-4-en-8-yl N-(4-methylbenzenesulfonyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
347.44
MDL number:

Iupac Name

(3aR,4S,5R,7S,7aR)-3a,4,5,6,7,7a-hexahydro-1H-4,7-methanoinden-5-yl tosylcarbamate

Inchi Code

1S/C18H21NO4S/c1-11-5-7-13(8-6-11)24(21,22)19-18(20)23-17-10-12-9-16(17)15-4-2-3-14(12)15/h2,4-8,12,14-17H,3,9-10H2,1H3,(H,19,20)

InChI key

IABLGVHYLVGUFH-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Gum

Product Link

https://www.keyorganics.net/sku/MS-3521

pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/MS-3521


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service