KEY465194558
Key Organics / BIONET
N-({4-[4-(hydroxymethyl)piperidin-1-yl]phenyl}methyl)-4-methylbenzene-1-sulfonamide
Select a Size
Vendor SKU: 2W-0309-500MG
$595.00
Select a Size
About This Item
Vendor SKU: 2W-0309-500MG
$595.00
Iupac Name
N-{4-[4-(hydroxymethyl)-1-piperidinyl]benzyl}-4-methylbenzenesulfonamide
Inchi Code
1S/C20H26N2O3S/c1-16-2-8-20(9-3-16)26(24,25)21-14-17-4-6-19(7-5-17)22-12-10-18(15-23)11-13-22/h2-9,18,21,23H,10-15H2,1H3
InChI key
PABCTOMGZLXKRI-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Product Link
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/2W-0309
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service