KEY465193551
Key Organics / BIONET
2-bromo-2-[(4-chlorophenyl)sulfanyl]-1-phenylethan-1-one
Select a Size
Vendor SKU: 1T-0009-500MG
$595.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
Vendor SKU: 1T-0009-500MG
$595.00
Notify Me
Get notified when this item is ready to ship via email.
Iupac Name
2-bromo-2-[(4-chlorophenyl)sulfanyl]-1-phenylethanone
Inchi Code
1S/C14H10BrClOS/c15-14(13(17)10-4-2-1-3-5-10)18-12-8-6-11(16)7-9-12/h1-9,14H
InChI key
VCFSMHYSFPUNEV-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Product Link
Signal Word
Danger
Hazard Codes
Precautionary statements
MSDS
https://www.keyorganics.net/sds/1T-0009
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service