KEY465193421
Key Organics / BIONET
ethyl (3E)-3-[(2-cyanoacetamido)imino]hexanoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
102 -103
Iupac Name
ethyl (3E)-3-[(cyanoacetyl)hydrazono]hexanoate
Inchi Code
1S/C11H17N3O3/c1-3-5-9(8-11(16)17-4-2)13-14-10(15)6-7-12/h5,13H,3-4,6,8H2,1-2H3,(H,14,15)
InChI key
JKCKRRSQLSJTMY-UHFFFAOYSA-N
Purity
90%
Country of Origin
GB
Physical Form
Gum
Product Link
https://www.keyorganics.net/sku/1R-0108
Melting Point
102 -103
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/1R-0108
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service