Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY448422204

Key Organics / BIONET

1,3-diethyl 2-[(dimethylamino)methylidene]propanedioate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$286.25
50 G
$556.25

Vendor SKU: STR09710-25G

$286.25


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
25 G
$286.25
50 G
$556.25

About This Item

CAS Number:
Molecular Weight:
215.25
MDL number:

Vendor SKU: STR09710-25G

$286.25


Ships in 21 days - from Aldrich Partner.

Iupac Name

diethyl 2-[(dimethylamino)methylene]malonate

Inchi Code

1S/C10H17NO4/c1-5-14-9(12)8(7-11(3)4)10(13)15-6-2/h7H,5-6H2,1-4H3

InChI key

XNSXXTKUKYWLAH-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Liquid,Solid

Product Link

https://www.keyorganics.net/sku/STR09710

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service