KEY432442015
Key Organics / BIONET
2-chloro-N-(2-methoxy-5-methylphenyl)acetamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
82 - 84
Iupac Name
2-chloro-N-(2-methoxy-5-methylphenyl)acetamide
Inchi Code
1S/C10H12ClNO2/c1-7-3-4-9(14-2)8(5-7)12-10(13)6-11/h3-5H,6H2,1-2H3,(H,12,13)
InChI key
ZVVBFKIUBBHWEV-UHFFFAOYSA-N
Purity
97%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/JS-2984
Melting Point
82 - 84
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/JS-2984
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service