Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY432310514

Key Organics / BIONET

(1E)-2-[4-(trifluoromethyl)phenyl]ethene-1-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
270.66
MDL number:

bp

356 - 357

mp

96 - 98

Iupac Name

(E)-2-(4-(trifluoromethyl)phenyl)ethene-1-sulfonyl chloride

Inchi Code

1S/C9H6ClF3O2S/c10-16(14,15)6-5-7-1-3-8(4-2-7)9(11,12)13/h1-6H/b6-5+

InChI key

REBMFKYPZPZKKL-AATRIKPKSA-N

storage temp.

Store under inert gas

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/AS-7063

Boiling point

356 - 357

Melting Point

96 - 98

pictograms

CorrosionExclamation mark

Signal Word

Danger

MSDS

https://www.keyorganics.net/sds/AS-7063


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service