KEY424062572
Key Organics / BIONET
benzyl 9-oxo-3-azaspiro[5.5]undecane-3-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
benzyl 9-oxo-3-azaspiro[5.5]undecane-3-carboxylate
Inchi Code
1S/C18H23NO3/c20-16-6-8-18(9-7-16)10-12-19(13-11-18)17(21)22-14-15-4-2-1-3-5-15/h1-5H,6-14H2
InChI key
XWVRAUFBYDPEPY-UHFFFAOYSA-N
Purity
95%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/SS-4793
Signal Word
Warning
Hazard Codes
Precautionary statements
MSDS
https://www.keyorganics.net/sds/SS-4793
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service