KEY424062567
Key Organics / BIONET
(2R,3R,4S,5R)-2-[6-amino-2-(propylsulfanyl)-9H-purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
About This Item
Iupac Name
(2R,3R,4S,5R)-2-(6-amino-2-(propylthio)-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
Inchi Code
1S/C13H19N5O4S/c1-2-3-23-13-16-10(14)7-11(17-13)18(5-15-7)12-9(21)8(20)6(4-19)22-12/h5-6,8-9,12,19-21H,2-4H2,1H3,(H2,14,16,17)/t6-,8-,9-,12-/m1/s1
InChI key
RZECJIGAYNYQAD-WOUKDFQISA-N
Purity
Country of Origin
Physical Form
Product Link
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/SS-4786
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service