Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY182144901

Key Organics / BIONET

ethyl 3-(chlorosulfonyl)propanoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
200.64
MDL number:

Iupac Name

ethyl 3-(chlorosulfonyl)propanoate

Inchi Code

1S/C5H9ClO4S/c1-2-10-5(7)3-4-11(6,8)9/h2-4H2,1H3

InChI key

JIQDEHYHHKZBRI-UHFFFAOYSA-N

storage temp.

Store under inert gas at 4C

Purity

85%

Country of Origin

GB

Physical Form

Liquid

Product Link

https://www.keyorganics.net/sku/GC-0767

pictograms

CorrosionExclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/GC-0767


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service