Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY181740425

Key Organics / BIONET

4-hydroxy-2-(pyridin-2-yl)pyrimidine-5-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
217.18
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

330 dec

Iupac Name

6-oxo-2-(2-pyridinyl)-1,6-dihydro-5-pyrimidinecarboxylic acid

Inchi Code

1S/C10H7N3O3/c14-9-6(10(15)16)5-12-8(13-9)7-3-1-2-4-11-7/h1-5H,(H,15,16)(H,12,13,14)

InChI key

XIXAYTYRVOIVMM-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/GC-0765

Melting Point

330 dec

pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary statements

MSDS

https://www.keyorganics.net/sds/GC-0765


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service