Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY128004454

Key Organics / BIONET

2-chloro-N-(4-methylphenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
183.64
MDL number:

mp

164 - 166

Iupac Name

2-chloro-N-(4-methylphenyl)acetamide

Inchi Code

1S/C9H10ClNO/c1-7-2-4-8(5-3-7)11-9(12)6-10/h2-5H,6H2,1H3,(H,11,12)

InChI key

CITIOELQTFSEGI-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/JS-198C

Melting Point

164 - 166

Signal Word

Danger

MSDS

https://www.keyorganics.net/sds/JS-198C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service