Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY128004274

Key Organics / BIONET

{4H,5H,6H-cyclopenta[b]thiophen-2-yl}methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
154.23
MDL number:

bp

48 - 54C @ 1mBar

Iupac Name

5,6-dihydro-4H-cyclopenta[b]thien-2-ylmethanol

Inchi Code

1S/C8H10OS/c9-5-7-4-6-2-1-3-8(6)10-7/h4,9H,1-3,5H2

InChI key

MOARKVWJFPIWFD-UHFFFAOYSA-N

storage temp.

Store under inert gas at 4C, Hygroscopic

Purity

90%

Country of Origin

GB

Physical Form

Liquid,Low Melting Solid

Product Link

https://www.keyorganics.net/sku/BB-0827

Boiling point

48 - 54C @ 1mBar

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service