KEY013018309
Key Organics / BIONET
ethyl 4-methyl-2-(pyridin-2-yl)-1,3-thiazole-5-carboxylate
Select a Size
Vendor SKU: 2G-303S-500MG
$167.50
Select a Size
About This Item
Vendor SKU: 2G-303S-500MG
$167.50
mp
82 - 84
Iupac Name
ethyl 4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate
Inchi Code
1S/C12H12N2O2S/c1-3-16-12(15)10-8(2)14-11(17-10)9-6-4-5-7-13-9/h4-7H,3H2,1-2H3
Inchi Key
UGCVLLXXBVTJPK-UHFFFAOYSA-N
Purity
Country of Origin
Physical Form
Product Link
Melting Point
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/2G-303S
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service