Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY007387644

Key Organics / BIONET

7-benzyl-4-chloro-5,6-diphenyl-7H-pyrrolo[2,3-d]pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
395.89
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

190 - 192

Iupac Name

7-benzyl-4-chloro-5,6-diphenyl-7H-pyrrolo[2,3-d]pyrimidine

Inchi Code

1S/C25H18ClN3/c26-24-22-21(19-12-6-2-7-13-19)23(20-14-8-3-9-15-20)29(25(22)28-17-27-24)16-18-10-4-1-5-11-18/h1-15,17H,16H2

InChI key

CLNNUPXMPYFDLW-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/9L-352S

Melting Point

190 - 192

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/9L-352S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service