Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY004181316

Key Organics / BIONET

1-chloro-3,4-dihydronaphthalene-2-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
192.64
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

36 - 39

Iupac Name

1-chloro-3,4-dihydro-2-naphthalenecarbaldehyde

Inchi Code

1S/C11H9ClO/c12-11-9(7-13)6-5-8-3-1-2-4-10(8)11/h1-4,7H,5-6H2

Inchi Key

GBRFPPIJEHQBGK-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/12N-501S

Melting Point

36 - 39

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/12N-501S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service