KEY004181239
Key Organics / BIONET
ethyl 4-chloro-2-phenylpyrimidine-5-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
130 - 131
Iupac Name
ethyl 4-chloro-2-phenyl-5-pyrimidinecarboxylate
Inchi Code
1S/C13H11ClN2O2/c1-2-18-13(17)10-8-15-12(16-11(10)14)9-6-4-3-5-7-9/h3-8H,2H2,1H3
InChI key
VXWYTPARKVGWFX-UHFFFAOYSA-N
storage temp.
Store under inert gas
Purity
95%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/10M-550S
Melting Point
130 - 131
Signal Word
Warning
Hazard Codes
Precautionary statements
MSDS
https://www.keyorganics.net/sds/10M-550S
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service