Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KEY001142133

Key Organics / BIONET

1-(6-chloropyridin-2-yl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
197.67
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

1-(6-chloro-2-pyridinyl)piperazine

Inchi Code

1S/C9H12ClN3/c10-8-2-1-3-9(12-8)13-6-4-11-5-7-13/h1-3,11H,4-7H2

InChI key

KQFRTJUIRPBBAB-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Low Melting Solid,Solid

Product Link

https://www.keyorganics.net/sku/6B-103

pictograms

Skull and crossbones

Signal Word

Danger

MSDS

https://www.keyorganics.net/sds/6B-103


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service