KEY001129506
Key Organics / BIONET
2-(2,4-dichlorophenoxy)benzaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
147C @ 0.1mm Hg
mp
58 - 60C
Iupac Name
2-(2,4-dichlorophenoxy)benzaldehyde
Inchi Code
1S/C13H8Cl2O2/c14-10-5-6-13(11(15)7-10)17-12-4-2-1-3-9(12)8-16/h1-8H
InChI key
AEAYPCKDEZKJMI-UHFFFAOYSA-N
Purity
95%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/12N-058
Boiling point
147C @ 0.1mm Hg
Melting Point
58 - 60C
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/12N-058
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service